Learn More
Tocris Bioscience™ GlyH 101
Reversible, voltage-dependent CFTR chloride channel blocker
Marca: Tocris Bioscience™ 5485/10
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
GlyH 101 is a reversible, voltage-dependent CFTR chloride channel blocker (Ki = 4.3 μM). Inhibits forskolin -induced hyperpolarization in nasal potential differences and inhibits cholera toxin-induced intestinal fluid secretion in mice.Especificaciones
| GlyH 101 | |
| N-2-Naphthalenyl-2-[(3,5-dibromo-2,4-dihydroxyphenyl)methylene]glycine hydrazide | |
| Soluble to 100mM in DMSO | |
| Bioactive Small Molecule |
| 328541-79-3 | |
| 10 mg | |
| Solid |
Título del producto
Research Use Only. Not For Human or Veterinary Use