CAS RN 31362-50-2
IUPAC Nombre:
N-{4-carbamimidamido-1-[(1-{[({2-carbamoyl-1-[(3-carbamoyl-1-{[1-({1-[(1-{[({1-[(1-{[1-carbamoyl-3-(methylsulfanyl)propyl]carbamoyl}-3-methylbutyl)carbamoyl]-2-(1H-imidazol-5-yl)ethyl}carbamoyl)methyl]carbamoyl}-2-methylpropyl)carbamoyl]ethyl}carbamoyl)-2-(1H-indol-3-yl)ethyl]carbamoyl}propyl)carbamoyl]ethyl}carbamoyl)methyl]carbamoyl}-3-methylbutyl)carbamoyl]butyl}-2-[(5-oxopyrrolidin-2-yl)formamido]pentanediamide
Sinónimos:
bombesin
Peso molecular (g/mol):
1619.87
Fórmula molecular:
C71H110N24O18S
InChi Key:
QXZBMSIDSOZZHK-UHFFFAOYNA-N
SMILES:
CSCCC(NC(=O)C(CC(C)C)NC(=O)C(CC1=CN=CN1)NC(=O)CNC(=O)C(NC(=O)C(C)NC(=O)C(CC1=CNC2=CC=CC=C12)NC(=O)C(CCC(N)=O)NC(=O)C(CC(N)=O)NC(=O)CNC(=O)C(CC(C)C)NC(=O)C(CCCNC(N)=N)NC(=O)C(CCC(N)=O)NC(=O)C1CCC(=O)N1)C(C)C)C(N)=O
1
–
4
de
4
resultados
1
MedChemExpress Bombesin
MedChemExpress Bombesin, a tetradecapeptide, plays an important role in the release of gastrin and the activation of G-protein receptors.
2
MedChemExpress Bombesin
MedChemExpress Bombesin, a tetradecapeptide, plays an important role in the release of gastrin and the activation of G-protein receptors.
3
MedChemExpress Bombesin
MedChemExpress Bombesin, a tetradecapeptide, plays an important role in the release of gastrin and the activation of G-protein receptors.
4
MedChemExpress Bombesin
MedChemExpress Bombesin, a tetradecapeptide, plays an important role in the release of gastrin and the activation of G-protein receptors.