CAS RN 30751-05-4
IUPAC Nombre:
3,4,5-trimethoxy-N-(piperidin-3-yl)benzamide
Sinónimos:
troxipide
Peso molecular (g/mol):
294.35
Fórmula molecular:
C15H22N2O4
InChi Key:
YSIITVVESCNIPR-UHFFFAOYNA-N
SMILES:
COC1=CC(=CC(OC)=C1OC)C(=O)NC1CCCNC1
1
–
2
de
2
resultados
1
Troxípide, TRC
CAS: 30751-05-4 Fórmula molecular: C15H22N2O4 Peso molecular (g/mol): 294.35 Sinónimo: KU 54,3-(3,4,5-Trimethoxybenzamido)piperidine,3,4,5-Trimethoxy-N-3-piperidinylbenzamide,Aplace Nombre IUPAC: 3,4,5-trimetoxi-N-piperidina-3-ilbenzamida SMILES: COc1cc(cc(OC)c1OC)C(=O)NC2CCCNC2
2
Troxipide, MedChemExpress
MedChemExpress Troxipide is an orally active defensive factor-enhancing therapeutic agent for gastritis and gastric ulcer (GU). Troxipide is a non-antisecretory gastro protective agent with antiulcer, anti-inflammatory and mucus-secreting properties.