CAS RN 304896-28-4
IUPAC Nombre:
2-cyano-3-[5-(2,5-dichlorophenyl)furan-2-yl]-N-(quinolin-5-yl)prop-2-enamide
Sinónimos:
2-cyano-3-[5-(2,5-dichlorophenyl)furan-2-yl]-N-(quinolin-5-yl)prop-2-enamide
Peso molecular (g/mol):
434.28
Fórmula molecular:
C23H13Cl2N3O2
InChi Key:
SVENPFFEMUOOGK-UHFFFAOYSA-N
SMILES:
ClC1=CC(C2=CC=C(O2)C=C(C#N)C(=O)NC2=C3C=CC=NC3=CC=C2)=C(Cl)C=C1
1
–
1
de
1
resultados
1
AGK2, MedChemExpress
MedChemExpress AGK2 is a selective SIRT2 inhibitor with an IC50 of 3.5 μM. AGK2 inhibits SIRT1 and SIRT3 with IC50s of 30 and 91 μM, respectively.