CAS RN 303052-45-1
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
2
de
2
resultados
1
MedChemExpress Neuropeptide Y(29-64)
MedChemExpress Neuropeptide Y(29-64) is a 36 amino acid peptide, a fragment of Neuropeptide Y.
2
MedChemExpress Neuropeptide Y(29-64)
MedChemExpress Neuropeptide Y(29-64) is a 36 amino acid peptide, a fragment of Neuropeptide Y.