CAS RN 301836-43-1
IUPAC Nombre:
4-[4-(2,3-dihydro-1,4-benzodioxin-6-yl)-5-(pyridin-2-yl)-1H-imidazol-2-yl]benzamide
Sinónimos:
4-[4-(2,3-dihydro-1,4-benzodioxin-6-yl)-5-(pyridin-2-yl)-1H-imidazol-2-yl]benzamide
Peso molecular (g/mol):
398.42
Fórmula molecular:
C23H18N4O3
InChi Key:
DPDZHVCKYBCJHW-UHFFFAOYSA-N
SMILES:
NC(=O)C1=CC=C(C=C1)C1=NC(=C(N1)C1=CC=CC=N1)C1=CC2=C(OCCO2)C=C1
1
–
3
de
3
resultados
1
Tocris Bioscience™ D 4476
CAS: 301836-43-1 Fórmula molecular: C23H18N4O3
2
D4476, MedChemExpress
MedChemExpress D4476 is a potent, selective and cell-permeable inhibitor of casein kinase 1(CK1) with an IC50 value of 0.3 μM in vitro.
3