accessibility menu, dialog, popup

UserName

PbCH3OOCH3OO

CAS RN 301-04-2

IUPAC Nombre: λ²-lead(2+) diacetate
Sinónimos: lead di(acetate) lead(II) acetate
Peso molecular (g/mol): 325.3
Fórmula molecular: C4H6O4Pb
InChi Key: GUWSLQUAAYEZAF-UHFFFAOYSA-L
SMILES: [Pb++].CC([O-])=O.CC([O-])=O

 No hay refinamientos