CAS RN 299-84-3
IUPAC Nombre:
O,O-dimethyl O-2,4,5-trichlorophenyl phosphorothioate
Sinónimos:
ronnel 芬氯磷 dermaphos caswell no. 724 C8H8cl3O3PS nanchor etrolene fenchlorphos korlan trichlormetaphos
Peso molecular (g/mol):
321.53
Fórmula molecular:
C8H8Cl3O3PS
InChi Key:
JHJOOSLFWRRSGU-UHFFFAOYSA-N
SMILES:
COP(=S)(OC)OC1=CC(Cl)=C(Cl)C=C1Cl
1
–
4
de
4
resultados
1
Fenchlorphos, MedChemExpress
MedChemExpress Fenchlorphos, an organophosphate, is an insecticide. Fenchlorphos is an inhibitor of the enzyme acetylcholinesterase (AChE). Fenchlorphos is able to cause mitochondrial dysfunction.