CAS RN 290297-26-6
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
2
de
2
resultados
1
Netupitant, TRC
CAS: 290297-26-6 Fórmula molecular: C30 H32 F6 N4 O Peso molecular (g/mol): 578.59 Sinónimo: N,α,α-Trimethyl-N-[4-(2-methylphenyl)-6-(4-methyl-1-piperazinyl)-3-pyridinyl]-3,5-bis(trifluoromethyl)-benzeneacetamide Nombre IUPAC: 2-[3,5-bis(trifluorometil)fenilo]-N,2-dimetil-N-[4-(2-metilfenil)-6-(4-metilpiperazina-1-il)piridina-3-il]propanamida SMILES: CN(C(=O)C(C)(C)c1cc(cc(c1)C(F)(F)F)C(F)(F)F)c2cnc(cc2c3ccccc3C)N4CCN(C)CC4
2
Netupitant, MedChemExpress
MedChemExpress Netupitant (CID-6451149) is a highly potent, selective and orally active neurokinin-1 (NK1) receptor antagonist with a Ki of 0.95 nM for hNK1 in CHO cells. Netupitant has antiemetic affect.