CAS RN 283609-79-0
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
3
de
3
resultados
1
MedChemExpress LSKL, Inhibitor of Thrombospondin (TSP-1)
MedChemExpress LSKL, Inhibitor of Thrombospondin (TSP-1) is a latency-associated protein (LAP)-TGFβ derived tetrapeptide and a competitive TGF-β1 antagonist.
2
MedChemExpress LSKL, Inhibitor of Thrombospondin (TSP-1)
MedChemExpress LSKL, Inhibitor of Thrombospondin (TSP-1) is a latency-associated protein (LAP)-TGFβ derived tetrapeptide and a competitive TGF-β1 antagonist.
3
MedChemExpress LSKL, Inhibitor of Thrombospondin (TSP-1)
MedChemExpress LSKL, Inhibitor of Thrombospondin (TSP-1) is a latency-associated protein (LAP)-TGFβ derived tetrapeptide and a competitive TGF-β1 antagonist.