Learn More
MedChemExpress LSKL, Inhibitor of Thrombospondin (TSP-1)
Código de producto. 16421958
Encompass_Preferred
Change view
Click to view available options
Cantidad:
5 mg
Tamaño de la unidad:
5 miligramos
16421958 5 mg 5 miligramos
1 options
Código de producto. 16421958
Marca: MedChemExpress HYP0299
MedChemExpress LSKL, Inhibitor of Thrombospondin (TSP-1) is a latency-associated protein (LAP)-TGFβ derived tetrapeptide and a competitive TGF-β1 antagonist.
LSKL, Inhibitor of Thrombospondin (TSP-1) inhibits the binding of TSP-1 to LAP and alleviates renal interstitial fibrosis and hepatic fibrosis. LSKL, Inhibitor of Thrombospondin (TSP-1) suppresses subarachnoid fibrosis via inhibition of TSP-1-mediated TGF-β1 activity, prevents the development of chronic hydrocephalus and improves long-term neurocognitive defects following subarachnoid hemorrhage (SAH). LSKL, Inhibitor of Thrombospondin (TSP-1) can readily crosse the blood-brain barrier.
Especificaciones
CAS 283609-79-0
Contenido y almacenamiento Please store the product under the recommended conditions in the Certificate of Analysis.
Para utilizar con (aplicación) Cancer-Kinase/protease
Peso molecular 458.6 g/mol
Tipo de producto Peptides
Cantidad 5 mg
Condiciones de envío Room temperature
Solubilidad DMSO : ≥ 100 mg/mL (218.05 mM) ∣H2O : 50 mg/mL (109.03 mM∣Need ultrasonic)