Learn More
MedChemExpress LSKL, Inhibitor of Thrombospondin (TSP-1)
Descripción
LSKL, Inhibitor of Thrombospondin (TSP-1) inhibits the binding of TSP-1 to LAP and alleviates renal interstitial fibrosis and hepatic fibrosis. LSKL, Inhibitor of Thrombospondin (TSP-1) suppresses subarachnoid fibrosis via inhibition of TSP-1-mediated TGF-β1 activity, prevents the development of chronic hydrocephalus and improves long-term neurocognitive defects following subarachnoid hemorrhage (SAH). LSKL, Inhibitor of Thrombospondin (TSP-1) can readily crosse the blood-brain barrier.
Especificaciones
Especificaciones
| CAS | 283609-79-0 |
| Contenido y almacenamiento | Please store the product under the recommended conditions in the Certificate of Analysis. |
| Para utilizar con (aplicación) | Cancer-Kinase/protease |
| Peso molecular | 458.6 g/mol |
| Tipo de producto | Peptides |
| Cantidad | 10 mg |
| Condiciones de envío | Room temperature |
| Solubilidad | DMSO : ≥ 100 mg/mL (218.05 mM) ∣H2O : 50 mg/mL (109.03 mM∣Need ultrasonic) |