CAS RN 264622-58-4
IUPAC Nombre:
N-(4-cyanophenyl)-2-[4-(2,6-dioxo-1,3-dipropyl-2,3,6,7-tetrahydro-1H-purin-8-yl)phenoxy]acetamide
Sinónimos:
N-(4-cyanophenyl)-2-[4-(2,6-dioxo-1,3-dipropyl-7H-purin-8-yl)phenoxy]acetamide
Peso molecular (g/mol):
486.53
Fórmula molecular:
C26H26N6O4
InChi Key:
AJBBEYXFRYFVNM-UHFFFAOYSA-N
SMILES:
CCCN1C2=C(NC(=N2)C2=CC=C(OCC(=O)NC3=CC=C(C=C3)C#N)C=C2)C(=O)N(CCC)C1=O
1
–
2
de
2
resultados
1
Tocris Bioscience™ MRS 1754
CAS: 264622-58-4 Fórmula molecular: C26H26N6O4
2
MRS 1754, MedChemExpress
MedChemExpress MRS 1754 is a selective antagonist radioligand for A2B adenosine receptor with very low affinity for A1 and A3 receptors of both humans and rats.