CAS RN 2444713-88-4
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Carboxy pyridostatin trifluoroacetate salt, MedChemExpress
MedChemExpress Carboxy pyridostatin trifluoroacetate salt has the peculiarity to exhibit high molecular specificity for RNA over DNA G4s.