CAS RN 2438721-48-1
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
T3-ATA (S-isomer), MedChemExpress
MedChemExpress T3-ATA S-isomer is the S-isomer of T3-ATA, which is the active form of the thyroid hormone.