CAS RN 2419-94-5
IUPAC Nombre:
(2S)-2-{[(tert-butoxy)carbonyl]amino}pentanedioate
Sinónimos:
(2S)-2-[(tert-butoxycarbonyl)amino]pentanedioate
Peso molecular (g/mol):
245.23
Fórmula molecular:
C10H15NO6
InChi Key:
AQTUACKQXJNHFQ-LURJTMIESA-L
SMILES:
CC(C)(C)OC(=O)N[C@@H](CCC([O-])=O)C([O-])=O
1
–
1
de
1
resultados
1
Ácido N-Boc-L-glutámico, 98 %, Thermo Scientific Chemicals
CAS: 2419-94-5 Fórmula molecular: C10H15NO6 Peso molecular (g/mol): 245.23 Número MDL: MFCD00037297 Clave InChI: AQTUACKQXJNHFQ-LURJTMIESA-L Sinónimo: boc-glu-oh,boc-l-glutamic acid,n-boc-l-glutamic acid,boc-l-glu-oh,n-tert-butoxycarbonyl-l-glutamic acid,n-t-butoxycarbonyl-l-glutamic acid,n-alpha-t-boc-l-glutamic acid,n-tert-butoxycarbonyl glutamic acid,l-glutamic acid, n-1,1-dimethylethoxy carbonyl,n-alpha-t-butyloxycarbonyl-l-glutamic acid PubChem CID: 2724758 SMILES: CC(C)(C)OC(=O)N[C@@H](CCC([O-])=O)C([O-])=O