CAS RN 23827-91-0
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
3
de
3
resultados
1
MedChemExpress [Des-Arg9]-Bradykinin acetate
MedChemExpress [Des-Arg9]-Bradykinin acetate is a Bradykinin B1 receptor agonist that displays selectivity for B1 over B2 receptors.
2
MedChemExpress [Des-Arg9]-Bradykinin acetate
MedChemExpress [Des-Arg9]-Bradykinin acetate is a Bradykinin B1 receptor agonist that displays selectivity for B1 over B2 receptors.
3
MedChemExpress [Des-Arg9]-Bradykinin acetate
MedChemExpress [Des-Arg9]-Bradykinin acetate is a Bradykinin B1 receptor agonist that displays selectivity for B1 over B2 receptors.