CAS RN 2375858-08-3
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Pyridostigmine-d6 bromide, MedChemExpress
MedChemExpress Pyridostigmine D6 bromide is the deuterium labeled Pyridostigmine, which is a parasympathomimetic and a reversible cholinesterase inhibitor.