CAS RN 23007-85-4
IUPAC Nombre:
hydrogen 1-methyl-4-phenyl-1,2,3,6-tetrahydropyridine chloride
Sinónimos:
hydrogen MPTP chloride
Peso molecular (g/mol):
209.72
Fórmula molecular:
C12H16ClN
InChi Key:
KOWJANGMTAZWDT-UHFFFAOYSA-N
SMILES:
[H+].[Cl-].CN1CCC(=CC1)C1=CC=CC=C1
1
–
1
de
1
resultados
1
MPTP hydrochloride, MedChemExpress
MedChemExpress MPTP hydrochloride is a brain penetrant dopamine neurotoxin, inducing Parkinson’s Disease. MPTP hydrochloride, a precusor of MPP+, induces apoptosis.