CAS RN 2271358-65-5
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
UAMC-3203 hydrochloride, MedChemExpress
MedChemExpress UAMC-3203 hydrochloride is a potent and selective Ferroptosis inhibitor with an IC50 of 12 nM.