CAS RN 226700-79-4
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Fosamprenavir, MedChemExpress
MedChemExpress Fosamprenavir (Amprenavir phosphate;GW 433908) is a phosphate ester prodrug of the antiretroviral protease inhibitor Amprenavir, with improved solubility. Anti-HIV infection.