CAS RN 2245794-64-1
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Tocris Bioscience™ Click N-Acetylmuramic acid - azide
Bacterial peptidoglycan derivative; suitable for 'click'-conjugation to fluorescent dyes