CAS RN 2239272-16-1
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
AWZ1066S, MedChemExpress
MedChemExpress AWZ1066S is a highly specific anti-Wolbachia drug candidate for a short-course treatment of filariasis, with an EC50 of 2.5 nM in cell assay.