CAS RN 2230849-55-3
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
EHMT2-IN-1, MedChemExpress
MedChemExpress EHMT2-IN-1 is a potent EHMT inhibitor, with IC50s of all <100 nM for EHMT1 peptide, EHMT2 peptide and cellular EHMT2. Used in the research of blood disorder or cancer.