CAS RN 220551-92-8
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
DAA-1106, MedChemExpress
MedChemExpress DAA1106 is a potent and selective ligand for peripheral benzodiazepine receptor (PBR), as a potent and selective agonist at the peripheral benzodiazepine receptor.