CAS RN 218924-25-5
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
KNK437, MedChemExpress
MedChemExpress KNK437 is a HSP inhibitor, and inhibits the induction of HSP105, HSP70, and HSP40.