CAS RN 21853-11-2
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
5α-Pregnane-3β,6α-diol-20-one, MedChemExpress
MedChemExpress 5α-Pregnane-3β,6α-diol-20-one is a mitogenic metabolite of progesterone, and it can be produced in starved androgen-responsive prostate cancer cells.