CAS RN 2183470-12-2
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
GCN2iB, MedChemExpress
MedChemExpress GCN2iB is an ATP-competitive inhibitor of a serine/threonine-protein kinase general control nonderepressible 2 (GCN2), with an IC50 of 2.4 nM.