CAS RN 218156-96-8
IUPAC Nombre:
N-[2-(piperidin-1-yl)-5-(trifluoromethyl)phenyl]pyridine-4-carboxamide
Sinónimos:
N-[2-(piperidin-1-yl)-5-(trifluoromethyl)phenyl]pyridine-4-carboxamide
Peso molecular (g/mol):
349.36
Fórmula molecular:
C18H18F3N3O
InChi Key:
DWFGGOFPIISJIT-UHFFFAOYSA-N
SMILES:
FC(F)(F)C1=CC(NC(=O)C2=CC=NC=C2)=C(C=C1)N1CCCCC1
1
–
1
de
1
resultados
1
SRPIN340, MedChemExpress
MedChemExpress SRPIN340 is an ATP-competitive serine-arginine-rich protein kinase (SRPK) inhibitor, with a Ki of 0.89 μM for SRPK1.