CAS RN 2173232-79-4
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
LOM612, MedChemExpress
MedChemExpress LOM612 is a potent FOXO relocator, with an EC50 value of 1.5 μM in U2fox RELOC cells.