CAS RN 215802-15-6
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
SB269652, MedChemExpress
MedChemExpress SB269652 is the first drug-like allosteric modulator of the dopamine D2 receptor (D2R); a new chemical probe that can differentiate D2R monomers from dimers or oligomers depending on the observed pharmacology.