CAS RN 2148339-28-8
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
2
de
2
resultados
1
Tocris Bioscience™ AS2717638
Selective, brain penetrant LPA5 antagonist
2
AS2717638, MedChemExpress
MedChemExpress AS2717638 is an oral active and selective lysophosphatidic acid receptor 5 (LPA5) antagonist, with an IC50 of 38 nM for hLPA5. AS2717638 also significantly improves PGE2-, PGF2α-, and AMPA-induced allodynia.