Learn More
AS2717638, MedChemExpress
Código de producto. 16484818
Encompass_Preferred
Cambiar vista
Click to view available options
Cantidad:
5 mg
10 mg
Tamaño de la unidad:
10 miligramos
5 miligramos
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
16484818 10 mg 10 miligramos
16477567 5 mg 5 miligramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Código de producto. 16484818 Proveedor Medchem Express N.º de proveedor HY114379
MedChemExpress AS2717638 is an oral active and selective lysophosphatidic acid receptor 5 (LPA5) antagonist, with an IC50 of 38 nM for hLPA5. AS2717638 also significantly improves PGE2-, PGF2α-, and AMPA-induced allodynia.
AS2717638 is an oral active and selective lysophosphatidic acid receptor 5 (LPA5) antagonist, with an IC50 of 38 nM for hLPA5. AS2717638 also significantly improves PGE2-, PGF2α-, and AMPA-induced allodynia.
Especificaciones
Nombre del producto químico o material AS2717638
CAS 2148339-28-8
Color Off-White
Notas de grado de pureza Research
Fórmula molecular C25H25N3O5
Cantidad 10 mg
Información de solubilidad DMSO : 60 mg/mL (134.08 mM; Need ultrasonic)
SMILES O=C1N(C2=NOC3=C2C=C(C)C=C3)C=C(C(N4CCCCC4)=O)C5=C1C=C(OC)C(OC)=C5
Peso molecular (g/mol) 447.48
Formula Weight (peso de la fórmula) 447.48
Porcentaje de pureza 98.07%
Grado Research
Forma física Powder
Para utilizar con (aplicación) Neuroscience-Neuromodulation