CAS RN 2140-77-4
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
N2-Methylguanosine, MedChemExpress
MedChemExpress N2-methylguanosine is a modified nucleoside that occurs at several specific locations in many tRNA's.