CAS RN 2140-69-4
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
2
de
2
resultados
1
3-Metiluridina, TRC
CAS: 2140-69-4 Fórmula molecular: C10H14N2O6 Peso molecular (g/mol): 258.23 Sinónimo: N3-Methyluridine,NSC 246077,3-Methyl-uridine SMILES: O[C@H]1[C@@H](O)[C@H](N2C(N(C)C(C=C2)=O)=O)O[C@@H]1CO
2
3-Methyluridine, MedChemExpress
MedChemExpress 3-Methyluridine (N3-Methyluridine) is a modified RNA nucleoside. 3-Methyluridine is often presents as RNA modification which can be detected in 23S rRNA of archaea, 16S and 23S rRNA of eubacteria, and 18S, 25S, and 28S of eukaryotic ribosomal RNAs.