CAS RN 2110449-60-8
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Glutaminyl Cyclase Inhibitor 1, MedChemExpress
MedChemExpress Glutaminyl Cyclase Inhibitor 1 is a glutaminyl cyclase inhibitor with an IC50 of 0.5 μM.