CAS RN 2098546-05-3
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
EBI-2511, MedChemExpress
MedChemExpress EBI-2511 is a highly potent and orally active EZH2 inhibitor, with an IC50 of 6 nM in Pfeffiera cell lines, respectively.