CAS RN 2097971-01-0
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
PROTAC BRD9 Degrader-1, MedChemExpress
MedChemExpress PROTAC BRD9 Degrader-1 is a PROTAC connected by ligands for Cereblon and BRD9 (IC50=13.5 nM), which can be used as a selective probe useful for the study of BAF complex biology.