Learn More
PROTAC BRD9 Degrader-1, MedChemExpress
Código de producto. 16486597
Encompass_Preferred
Cambiar vista
Click to view available options
Cantidad:
5 mg
10 mg
Tamaño de la unidad:
10 miligramos
5 miligramos
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
16486597 5 mg 5 miligramos
16433878 10 mg 10 miligramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Código de producto. 16486597 Proveedor Medchem Express N.º de proveedor HY103632
MedChemExpress PROTAC BRD9 Degrader-1 is a PROTAC connected by ligands for Cereblon and BRD9 (IC50=13.5 nM), which can be used as a selective probe useful for the study of BAF complex biology.
PROTAC BRD9 Degrader-1 is a PROTAC connected by ligands for Cereblon and BRD9 (IC50=13.5 nM), which can be used as a selective probe useful for the study of BAF complex biology.
Especificaciones
Nombre del producto químico o material PROTAC BRD9 Degrader-1
CAS 2097971-01-0
Color Yellow
Notas de grado de pureza Research
Fórmula molecular C42H45N7O12S2
Cantidad 5 mg
Información de solubilidad DMSO : 150 mg/mL (165.93 mM; Need ultrasonic)
SMILES O=C1C(C=C(C(NC(CC2)CCS2(=O)=O)=N)S3)=C3C(C4=CC(OC)=C(OCC(NCCCCNC(COC5=C(C(N6C(CCC7=O)C(N7)=O)=O)C(C6=O)=CC=C5)=O)=O)C=C4)=CN1C
Peso molecular (g/mol) 903.98
Formula Weight (peso de la fórmula) 903.98
Porcentaje de pureza 98.3%
Grado Research
Forma física Solid
Para utilizar con (aplicación) Neuroscience-Neuromodulation