CAS RN 2086772-26-9
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
JNJ-64619178, MedChemExpress
MedChemExpress JNJ-64619178 (Onametostat) is a selective, orally active and pseudo-irreversible protein arginine methyltransferase 5 (PRMT5) inhibitor with an IC50 of 0.14 nM. JNJ-64619178 has potent activity In lung cancer.