CAS RN 2070014-98-9
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
IFN alpha-IFNAR-IN-1 hydrochloride, MedChemExpress
MedChemExpress IFN alpha-IFNAR-IN-1 hydrochloride is a nonpeptidic, low-molecular-weight inhibitor of the interaction between IFN-α and IFNAR; inhibit MVA-induced IFN-α responses by BM-pDCs (IC50=2-8 uM).