CAS RN 2070009-54-8
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
NVP-CGM097 (stereoisomer), MedChemExpress
MedChemExpress NVP-CGM097 (stereoisomer) is a stereoisomer of NVP-CGM097, with no special bioactivity. NVP-CGM097 is a potent and selective MDM2 inhibitor.