CAS RN 2070009-26-4
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
3
de
3
resultados
1
MedChemExpress pGlu-Pro-Arg-MNA monoacetate
MedChemExpress pGlu-Pro-Arg-MNA monoacetate is a chromogenic substrate.
2
MedChemExpress pGlu-Pro-Arg-MNA monoacetate
MedChemExpress pGlu-Pro-Arg-MNA monoacetate is a chromogenic substrate.
3
MedChemExpress pGlu-Pro-Arg-MNA monoacetate
MedChemExpress pGlu-Pro-Arg-MNA monoacetate is a chromogenic substrate.