CAS RN 2061980-01-4
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
3
de
3
resultados
1
Tocris Bioscience™ XMU MP 1
Potent and selective ATP-competitive mammalian sterile 20-like kinase (MST) 1/2 inhibitor
2
Tocris Bioscience™ XMU MP 1
Potent and selective MST1/2 inhibitor
3
XMU-MP-1, MedChemExpress
MedChemExpress XMU-MP-1 is a reversible and selective MST1/2 inhibitor with IC50s of 71.1 and 38.1 nM, respectively.