CAS RN 203849-91-6
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
MMAD, MedChemExpress
MedChemExpress MMAD is a potent tubulin inhibitor, is a toxin payload in antibody drug conjugates (ADCs).