CAS RN 202057-76-9
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Manitimus, MedChemExpress
MedChemExpress Manitimus is an inhibitor of dehydroorotate dehydrogenase, and a potent immunosuppressive drug.