CAS RN 201608-14-2
IUPAC Nombre:
(4S)-4-{[(1S)-1-{[(1S)-2-carboxy-1-{[2-oxo-4-(trifluoromethyl)-2H-chromen-7-yl]carbamoyl}ethyl]carbamoyl}-2-methylpropyl]carbamoyl}-4-[(2S)-3-carboxy-2-acetamidopropanamido]butanoic acid
Sinónimos:
(4S)-4-{[(1S)-1-{[(1S)-2-carboxy-1-{[2-oxo-4-(trifluoromethyl)chromen-7-yl]carbamoyl}ethyl]carbamoyl}-2-methylpropyl]carbamoyl}-4-[(2S)-3-carboxy-2-acetamidopropanamido]butanoic acid
Peso molecular (g/mol):
729.62
Fórmula molecular:
C30H34F3N5O13
InChi Key:
GZDRODOYEFEHGG-NUDCOPPTSA-N
SMILES:
CC(C)[C@H](NC(=O)[C@H](CCC(O)=O)NC(=O)[C@H](CC(O)=O)NC(C)=O)C(=O)N[C@@H](CC(O)=O)C(=O)NC1=CC=C2C(OC(=O)C=C2C(F)(F)F)=C1
1
–
4
de
4
resultados
1
MedChemExpress Ac-DEVD-AFC
MedChemExpress Ac-DEVD-AFC is a fluorogenic substrate (λex=400 nm, λem=530 nm).
2
MedChemExpress Ac-DEVD-AFC
MedChemExpress Ac-DEVD-AFC is a fluorogenic substrate (λex=400 nm, λem=530 nm).
3
MedChemExpress Ac-DEVD-AFC
MedChemExpress Ac-DEVD-AFC is a fluorogenic substrate (λex=400 nm, λem=530 nm).