Learn More
MedChemExpress Ac-DEVD-AFC
Product Code. 16432799
Encompass_Preferred
Change view
Click to view available options
Cantidad:
1 mg
Unit Size:
1 miligramo
Product Code. Cantidad unitSize
16432799 1 mg 1 miligramo
1 options
This item is not returnable. View return policy
Product Code. 16432799 Supplier MedChemExpress Supplier No. HYP1005
This item is not returnable. View return policy
MedChemExpress Ac-DEVD-AFC is a fluorogenic substrate (λex=400 nm, λem=530 nm).
Specifications
CAS 201608-14-2
Solubilidad DMSO : ≥ 50 mg/mL (68.53 mM) ∣H2O : < 0.1 mg/mL (insoluble)
Contenido y almacenamiento Protect from light∣Powder -80°C 2 years, -20°C 1 year∣In solvent : -80°C, 6 months∣-20°C, 1 month (protect from light)
Para utilizar con (aplicación) Cancer-programmed cell death
Cantidad 1 mg
Peso molecular 729.61 g/mol
Tipo de producto Dye Reagents
Grado de pureza o calidad 99.15%