CAS RN 199807-33-5
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
2
de
2
resultados
1
MedChemExpress Cyclo(Arg-Gly-Asp-D-Phe-Val) TFA
MedChemExpress Cyclo(Arg-Gly-Asp-D-Phe-Val) (TFA) is an inhibitor of integrin αvβ3, with antitumor activity.
2
MedChemExpress Cyclo(Arg-Gly-Asp-D-Phe-Val) TFA
MedChemExpress Cyclo(Arg-Gly-Asp-D-Phe-Val) (TFA) is an inhibitor of integrin αvβ3, with antitumor activity.