CAS RN 199113-98-9
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Balaglitazone, MedChemExpress
MedChemExpress Balaglitazone is a selective partial PPARγ agonist with an EC50 of 1.351 μM for human PPARγ.